CAS RN 2142-67-8
IUPAC Name:
1-(2-ethoxyphenyl)ethan-1-one
Synonyms:
1-(2-ethoxyphenyl)ethanone
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
TVGMOUGXQYQZOL-UHFFFAOYSA-N
SMILES:
CCOC1=CC=CC=C1C(C)=O
1
–
1
of
1
results
1
2'-Ethoxyacetophenone 98.0+%, TCI America™
CAS: 2142-67-8 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00017262 InChI Key: TVGMOUGXQYQZOL-UHFFFAOYSA-N Synonym: 2'-ethoxyacetophenone,1-2-ethoxyphenyl ethanone,ethanone, 1-2-ethoxyphenyl,2-acetylphenetole,1-2-ethoxyphenyl ethan-1-one,1-acetyl-2-ethoxybenzene,o-ethoxyacetophenone,pubchem13191,acmc-1bt18,ethanone,2-ethoxy-1-phenyl PubChem CID: 578482 IUPAC Name: 1-(2-ethoxyphenyl)ethanone SMILES: CCOC1=CC=CC=C1C(=O)C