accessibility menu, dialog, popup

UserName

CAS RN 2136-75-6

IUPAC Name: 2-(triphenyl-λ⁵-phosphanylidene)acetaldehyde
Synonyms: 2-(triphenyl-λ⁵-phosphanylidene)acetaldehyde
Molecular Weight (g/mol): 304.33
Molecular Formula: C20H17OP
InChi Key: CQCAYWAIRTVXIY-UHFFFAOYSA-N
SMILES: O=CC=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (5)
  • (3)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)