accessibility menu, dialog, popup

UserName

H3CSiCH3H3CSiCH3CH3H3C

CAS RN 2117-28-4

IUPAC Name: trimethyl[(trimethylsilyl)methyl]silane
Synonyms: trimethyl[(trimethylsilyl)methyl]silane
Molecular Weight (g/mol): 160.41
Molecular Formula: C7H20Si2
InChi Key: GYIODRUWWNNGPI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C[Si](C)(C)C
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results