CAS RN 21168-41-2
IUPAC Name:
ethyl 4,6-dichloroquinoline-3-carboxylate
Synonyms:
ethyl 4,6-dichloroquinoline-3-carboxylate
Molecular Weight (g/mol):
270.11
Molecular Formula:
C12H9Cl2NO2
InChi Key:
BSSNTDZRVNQGDF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=CN=C2C=CC(Cl)=CC2=C1Cl
1
–
1
of
1
results
1
Promotions Available
Ethyl 4,6-dichloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 4,6-dichloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals
CAS: 21168-41-2 Molecular Formula: C12H9Cl2NO2 Molecular Weight (g/mol): 270.11 MDL Number: MFCD00173348 InChI Key: BSSNTDZRVNQGDF-UHFFFAOYSA-N Synonym: ethyl 4,6-dichloro-3-quinolinecarboxylate,4,6-dichloroquinoline-3-carboxylic acid ethyl ester,ethyl-4,6-dichloroquinoline-3-carboxylate,3-quinolinecarboxylic acid, 4,6-dichloro-, ethyl ester,ethyl-4, 6-dichloroquinoline-3-carboxylate,4,6-dichloro-3-quinolinecarboxylic acid ethyl ester,3-quinolinecarboxylicacid, 4,6-dichloro-, ethyl ester,4,6-dichloroquinoline-3-carboxylic acid, ethyl ester PubChem CID: 1479087 IUPAC Name: ethyl 4,6-dichloroquinoline-3-carboxylate SMILES: CCOC(=O)C1=CN=C2C=CC(Cl)=CC2=C1Cl