accessibility menu, dialog, popup

UserName

CH3OCH3O

CAS RN 2114-33-2

IUPAC Name: 1-phenylpropan-2-yl acetate
Synonyms: methylbenzyl carbinyl acetate
Molecular Weight (g/mol): 178.23
Molecular Formula: C11H14O2
InChi Key: LOHDWLZDQXQLJA-UHFFFAOYNA-N
SMILES: CC(CC1=CC=CC=C1)OC(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results