accessibility menu, dialog, popup

UserName

CH3OOOHOH

CAS RN 2107-76-8

IUPAC Name: 5,7-dihydroxy-4-methyl-2H-chromen-2-one
Synonyms: 5,7-dihydroxy-4-methylchromen-2-one
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: QNVWGEJMXOQQPM-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC(O)=CC(O)=C12
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)