accessibility menu, dialog, popup

UserName

CAS RN 2078-54-8

IUPAC Name: 2,6-bis(propan-2-yl)phenol
Synonyms: propofol propofol fresenius abbott brand of propofol rapinovet pisa brand of propofol fresofol propofol (substance) propofol preparation disoprofol parnell brand of propofol
Molecular Weight (g/mol): 178.28
Molecular Formula: C12H18O
InChi Key: OLBCVFGFOZPWHH-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC(C(C)C)=C1O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results