CAS RN 20776-48-1
IUPAC Name:
2-amino-6-bromobenzoic acid
Synonyms:
2-amino-6-bromobenzoic acid
Molecular Weight (g/mol):
216.03
Molecular Formula:
C7H6BrNO2
InChi Key:
BNQPROAXWQCNKO-UHFFFAOYSA-N
SMILES:
NC1=C(C(O)=O)C(Br)=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Amino-6-bromobenzoic acid, 97+%
2-Amino-6-bromobenzoic acid, 97+%
CAS: 20776-48-1 Molecular Formula: C7H6BrNO2 Molecular Weight (g/mol): 216.03 MDL Number: MFCD03618455 InChI Key: BNQPROAXWQCNKO-UHFFFAOYSA-N Synonym: 6-bromoanthranilic acid,3-bromo-2-carboxyaniline,benzoic acid, 2-amino-6-bromo,buttpark 49\07-50,2-amino-6-bromo-benzoic acid,ksc201s8r PubChem CID: 10560649 IUPAC Name: 2-amino-6-bromobenzoic acid SMILES: NC1=C(C(O)=O)C(Br)=CC=C1