CAS RN 207346-42-7
IUPAC Name:
methyl 2-amino-4,5-difluorobenzoate
Synonyms:
methyl 2-amino-4,5-difluorobenzoate
Molecular Weight (g/mol):
187.15
Molecular Formula:
C8H7F2NO2
InChi Key:
VIAQNTRKUPBQKR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(F)=C(F)C=C1N
1
–
1
of
1
results
1
Promotions Available
Methyl 2-amino-4,5-difluorobenzoate, 98%
Methyl 2-amino-4,5-difluorobenzoate, 98%
CAS: 207346-42-7 Molecular Formula: C8H7F2NO2 Molecular Weight (g/mol): 187.15 MDL Number: MFCD09264511 InChI Key: VIAQNTRKUPBQKR-UHFFFAOYSA-N Synonym: 2-amino-4,5-difluorobenzoic acid methyl ester,benzoic acid, 2-amino-4,5-difluoro-, methyl ester,pubchem4676,acmc-209fdn,ksc496g7j,methyl 4,5-difluoroanthranilate,methyl2-amino-4,5-difluorobenzoate,methyl 2-amino-4,5-difluoro-benzoate,2-amino-4,5-difluoro-benzoic acid methyl ester PubChem CID: 18472001 IUPAC Name: methyl 2-amino-4,5-difluorobenzoate SMILES: COC(=O)C1=CC(F)=C(F)C=C1N