accessibility menu, dialog, popup

UserName

H3COSBOHHO

CAS RN 206551-43-1

IUPAC Name: (5-acetylthiophen-2-yl)boronic acid
Synonyms: 5-acetylthiophen-2-ylboronic acid
Molecular Weight (g/mol): 169.99
Molecular Formula: C6H7BO3S
InChi Key: DCNMATSPQKWETQ-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(S1)B(O)O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results