missing translation for 'onlineSavingsMsg'
Learn More

5-Acetyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A23645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A23645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A23645G Supplier TCI America Supplier No. A23645G

Specifications

Chemical Name or Material 5-Acetyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride)
CAS 206551-43-1
Molecular Formula C6H7BO3S
Quantity 5 g
MDL Number MFCD01075681
Synonym 5-acetylthiophene-2-boronic acid, 5-acetyl-2-thiopheneboronic acid, 5-acetyl-2-thienylboronic acid, 5-acetylthiophen-2-yl boronic acid, 5-acetyl-2-thienyl boronic acid, 5-acetylthiophen-2-boronic acid, boronic acid, 5-acetyl-2-thienyl, 2-acetylthien-5-yl boronic acid, 5-acetylthien-2-yl boronic acid, 1-5-dihydroxyboranyl thiophen-2-yl ethan-1-one
InChI Key DCNMATSPQKWETQ-UHFFFAOYSA-N
SMILES B(C1=CC=C(S1)C(=O)C)(O)O
IUPAC Name (5-acetylthiophen-2-yl)boronic acid
Molecular Weight (g/mol) 169.989
PubChem CID 4143377
Formula Weight 169.99
Physical Form Crystalline Powder
Show More Show Less