accessibility menu, dialog, popup

UserName

CH3CH3CH3N

CAS RN 20654-47-1

IUPAC Name: 2,2-dimethylpentanenitrile
Synonyms: 2,2-dimethylpentanenitrile
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: TZJQCUDHKUWEFU-UHFFFAOYSA-N
SMILES: CCCC(C)(C)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results