accessibility menu, dialog, popup

UserName

CAS RN 20638-07-7

HONH2OHH2N

CAS RN 20638-07-7

IUPAC Name: 2-amino-4-[9-(3-amino-4-hydroxyphenyl)-9H-fluoren-9-yl]phenol
Synonyms: 2-amino-4-[9-(3-amino-4-hydroxyphenyl)fluoren-9-yl]phenol
Molecular Weight (g/mol): 380.45
Molecular Formula: C25H20N2O2
InChi Key: NLGOBIIKXFNGQR-UHFFFAOYSA-N
SMILES: NC1=C(O)C=CC(=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(N)=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results