accessibility menu, dialog, popup

UserName

OONH2NH2

CAS RN 20610-77-9

IUPAC Name: 4-aminophenyl 4-aminobenzoate
Synonyms: 4-aminophenyl 4-aminobenzoate
Molecular Weight (g/mol): 228.25
Molecular Formula: C13H12N2O2
InChi Key: LOCTYHIHNCOYJZ-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(=O)C2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results