accessibility menu, dialog, popup

UserName

CAS RN 204320-51-4

IUPAC Name: 4-{[(9H-fluoren-9-yl)methoxy]carbonyl}morpholine-3-carboxylic acid
Synonyms: 4-[(9H-fluoren-9-ylmethoxy)carbonyl]morpholine-3-carboxylic acid
Molecular Weight (g/mol): 353.37
Molecular Formula: C20H19NO5
InChi Key: CJVIYWXADATNKP-UHFFFAOYNA-N
SMILES: OC(=O)C1COCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results