accessibility menu, dialog, popup

UserName

OHFCl

CAS RN 2040-90-6

IUPAC Name: 2-chloro-6-fluorophenol
Synonyms: 2-chloro-6-fluoro-phenol
Molecular Weight (g/mol): 146.55
Molecular Formula: C6H4ClFO
InChi Key: QIAQIYQASAWZPP-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC=C1Cl
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results