accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 2033-76-3

IUPAC Name: 1-(2-bromoethoxy)-4-chlorobenzene
Synonyms: 1-(2-bromoethoxy)-4-chlorobenzene
Molecular Weight (g/mol): 235.51
Molecular Formula: C8H8BrClO
InChi Key: YYFLBDSMQRWARK-UHFFFAOYSA-N
SMILES: ClC1=CC=C(OCCBr)C=C1
Boiling Point 
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)