CAS RN 202865-66-5
CAS RN 202865-66-5
IUPAC Name:
(2-bromo-5-fluorophenyl)methanol
Synonyms:
(2-bromo-5-fluorophenyl)methanol
Molecular Weight (g/mol):
205.03
Molecular Formula:
C7H6BrFO
InChi Key:
HXGZPMHPSBJMKB-UHFFFAOYSA-N
SMILES:
OCC1=C(Br)C=CC(F)=C1
1
–
1
of
1
results
1
2-Bromo-5-fluorobenzyl alcohol, 97%, Thermo Scientific Chemicals
CAS: 202865-66-5 Molecular Formula: C7H6BrFO Molecular Weight (g/mol): 205.03 MDL Number: MFCD00142876 InChI Key: HXGZPMHPSBJMKB-UHFFFAOYSA-N Synonym: 2-bromo-5-fluorobenzyl alcohol,2-bromo-5-fluorophenyl methanol,2-bromo-5-fluorobenzylalcohol,benzenemethanol, 2-bromo-5-fluoro,2-bromo-5-fluorophenyl methan-1-ol,buttpark 87\01-69,2-bromo-5-fluorbenzyl alcohol,pubchem1963,acmc-1cs63,bromo-5-fluorobenzyl alcohol PubChem CID: 2773349 IUPAC Name: (2-bromo-5-fluorophenyl)methanol SMILES: OCC1=C(Br)C=CC(F)=C1