accessibility menu, dialog, popup

UserName

CAS RN 20170-32-5

IUPAC Name: 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Synonyms: 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 277.39
Molecular Formula: C17H25O3
InChi Key: WPMYUUITDBHVQZ-UHFFFAOYSA-M
SMILES: CC(C)(C)C1=CC(CCC([O-])=O)=CC(=C1O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results