accessibility menu, dialog, popup

UserName

FClBr

CAS RN 1996-29-8

IUPAC Name: 1-bromo-4-chloro-2-fluorobenzene
Synonyms: 1-bromo-4-chloro-2-fluorobenzene
Molecular Weight (g/mol): 209.44
Molecular Formula: C6H3BrClF
InChi Key: FPNVMCMDWZNTEU-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CC=C1Br
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)