accessibility menu, dialog, popup

UserName

CAS RN 19953-58-3

IUPAC Name: 2-methyl-6-phenyl-3H,7H-imidazo[1,2-a]pyrazin-3-one
Synonyms: 2-methyl-6-phenyl-7H-imidazo[1,2-a]pyrazin-3-one
Molecular Weight (g/mol): 225.25
Molecular Formula: C13H11N3O
InChi Key: JYQHFNUMLIRXCQ-UHFFFAOYSA-N
SMILES: CC1=NC2=CNC(=CN2C1=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results