accessibility menu, dialog, popup

UserName

CAS RN 198633-76-0

CH3FN

CAS RN 198633-76-0

IUPAC Name: 2-fluoro-6-methylbenzonitrile
Synonyms: 2-fluoro-6-methylbenzonitrile
Molecular Weight (g/mol): 135.14
Molecular Formula: C8H6FN
InChi Key: UCSKOUQDVWADGZ-UHFFFAOYSA-N
SMILES: CC1=C(C#N)C(F)=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results