accessibility menu, dialog, popup

UserName

HONOH

CAS RN 19780-85-9

IUPAC Name: 3-[1-(2-hydroxyethyl)piperidin-4-yl]propan-1-ol
Synonyms: 3-[1-(2-hydroxyethyl)piperidin-4-yl]propan-1-ol
Molecular Weight (g/mol): 187.28
Molecular Formula: C10H21NO2
InChi Key: QCZXNEGHMZUDNS-UHFFFAOYSA-N
SMILES: OCCCC1CCN(CCO)CC1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results