accessibility menu, dialog, popup

UserName

CAS RN 1975-50-4

CH3NOOOHO

CAS RN 1975-50-4

IUPAC Name: 2-methyl-3-nitrobenzoic acid
Synonyms: benzoic acid,2-methyl-3-nitro
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: YPQAFWHSMWWPLX-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1[N+]([O-])=O)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results