accessibility menu, dialog, popup

UserName

CH3OOH2NNH2

CAS RN 1949-55-9

IUPAC Name: methyl 3,5-diaminobenzoate
Synonyms: methyl 3,5-diaminobenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: IYWLDOMAEQHKJZ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(N)=CC(N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results