accessibility menu, dialog, popup

UserName

CAS RN 19471-12-6

NH2NH2

CAS RN 19471-12-6

IUPAC Name: 3-[(3-aminophenyl)methyl]aniline
Synonyms: 3-[(3-aminophenyl)methyl]aniline
Molecular Weight (g/mol): 198.27
Molecular Formula: C13H14N2
InChi Key: CKOFBUUFHALZGK-UHFFFAOYSA-N
SMILES: NC1=CC(CC2=CC(N)=CC=C2)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results