CAS RN 19408-46-9
IUPAC Name:
hydrogen {N'-[(2R,3S,5S,6R)-5-amino-2-methyl-6-{[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxy}oxan-3-yl]carbamimidoyl}formic acid chloride
Synonyms:
kasugamycin
Molecular Weight (g/mol):
415.82
Molecular Formula:
C14H26ClN3O9
InChi Key:
ZDRBJJNXJOSCLR-YZKQBBCCSA-N
SMILES:
[H+].[Cl-].C[C@H]1O[C@H](OC2[C@@H](O)[C@@H](O)C(O)[C@H](O)[C@H]2O)[C@@H](N)C[C@@H]1N=C(N)C(O)=O
1
–
1
of
1
results
1
Kasugamycin, 50mg/mL in distilled water, sterile-filtered, Thermo Scientific™
CAS: 19408-46-9 Molecular Formula: C14H26ClN3O9 Molecular Weight (g/mol): 415.824 MDL Number: MFCD00058459 InChI Key: ZDRBJJNXJOSCLR-YZKQBBCCSA-N PubChem CID: 66765861 IUPAC Name: 2-amino-2-[(2R,3S,5S,6R)-5-amino-2-methyl-6-[(2R,3S,5S,6S)-2,3,4,5,6-pentahydroxycyclohexyl]oxyoxan-3-yl]iminoacetic acid;hydron;chloride SMILES: [H+].CC1C(CC(C(O1)OC2C(C(C(C(C2O)O)O)O)O)N)N=C(C(=O)O)N.[Cl-]