accessibility menu, dialog, popup

UserName

CAS RN 1928-38-7

H3COOOClCl

CAS RN 1928-38-7

IUPAC Name: methyl 2-(2,4-dichlorophenoxy)acetate
Synonyms: 2,4-D, methyl ester
Molecular Weight (g/mol): 235.06
Molecular Formula: C9H8Cl2O3
InChi Key: HWIGZMADSFQMOI-UHFFFAOYSA-N
SMILES: COC(=O)COC1=CC=C(Cl)C=C1Cl
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)