accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 18979-53-8

IUPAC Name: 4-(pentyloxy)phenol
Synonyms: phenol, 4-(pentyloxy)-
Molecular Weight (g/mol): 180.25
Molecular Formula: C11H16O2
InChi Key: JCLFHZLOKITRCE-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results