accessibility menu, dialog, popup

UserName

CAS RN 189681-04-7

IUPAC Name: 4-chloro-5-(thiophen-2-yl)thieno[2,3-d]pyrimidine
Synonyms: 4-chloro-5-(thiophen-2-yl)thieno[2,3-d]pyrimidine
Molecular Weight (g/mol): 252.73
Molecular Formula: C10H5ClN2S2
InChi Key: VNWBUFWVNCUUKY-UHFFFAOYSA-N
SMILES: ClC1=C2C(SC=C2C2=CC=CS2)=NC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results