accessibility menu, dialog, popup

UserName

H3COFN

CAS RN 189628-38-4

IUPAC Name: 5-fluoro-2-methoxybenzonitrile
Synonyms: 5-fluoro-2-methoxybenzonitrile
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: XNCVSNKXADJGOG-UHFFFAOYSA-N
SMILES: COC1=C(C=C(F)C=C1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results