accessibility menu, dialog, popup

UserName

CH3NNOO

CAS RN 1885-76-3

IUPAC Name: 2-methyl-6-nitrobenzonitrile
Synonyms: 2-methyl-6-nitrobenzonitrile
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: BOTPDHNZLRJZOO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1C#N)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results