accessibility menu, dialog, popup

UserName

OOOI

CAS RN 1878-94-0

IUPAC Name: 2-(4-iodophenoxy)acetate
Synonyms: 2-(4-iodophenoxy)acetate
Molecular Weight (g/mol): 277.04
Molecular Formula: C8H6IO3
InChi Key: RKORKXFKXYYHAQ-UHFFFAOYSA-M
SMILES: [O-]C(=O)COC1=CC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results