accessibility menu, dialog, popup

UserName

CAS RN 1878-88-2

OHOONOO

CAS RN 1878-88-2

IUPAC Name: 2-(3-nitrophenoxy)acetic acid
Synonyms: M-nitrophenoxyacetic acid
Molecular Weight (g/mol): 197.15
Molecular Formula: C8H7NO5
InChi Key: BNRRQAASFDGMMQ-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results