accessibility menu, dialog, popup

UserName

CH3OOOHO

CAS RN 1878-80-4

IUPAC Name: 2-(3-acetylphenoxy)acetic acid
Synonyms: M-acetylphenoxyacetic acid
Molecular Weight (g/mol): 194.19
Molecular Formula: C10H10O4
InChi Key: VDZJGFDUSYCVJA-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(OCC(O)=O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results