accessibility menu, dialog, popup

UserName

CAS RN 187148-77-2

H3CH3CN

CAS RN 187148-77-2

IUPAC Name: 9-(2-ethylhexyl)-9H-carbazole
Synonyms: 9-(2-ethylhexyl)carbazole
Molecular Weight (g/mol): 279.43
Molecular Formula: C20H25N
InChi Key: SAXNQWFLHXTRDI-UHFFFAOYNA-N
SMILES: CCCCC(CC)CN1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results