missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(2-Ethylhexyl)carbazole 97.0+%, TCI America™
Specifications
Specifications
| CAS | 187148-77-2 |
| Color | Yellow |
| Molecular Formula | C20H25N |
| Quantity | 1 g |
| MDL Number | MFCD06798127 |
| InChI Key | SAXNQWFLHXTRDI-UHFFFAOYSA-N |
| SMILES | CCCCC(CC)CN1C2=CC=CC=C2C3=CC=CC=C31 |
| IUPAC Name | 9-(2-ethylhexyl)carbazole |
| Molecular Weight (g/mol) | 279.427 |
| PubChem CID | 16217868 |
| Show More |
Safety and Handling
| TSCA | No |