accessibility menu, dialog, popup

UserName

FFFSOOH2N

CAS RN 1869-24-5

IUPAC Name: 2-(trifluoromethyl)benzene-1-sulfonamide
Synonyms: 2-(trifluoromethyl)benzenesulfonamide
Molecular Weight (g/mol): 225.19
Molecular Formula: C7H6F3NO2S
InChi Key: AFFPZJFLSDVZBV-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CC=C1C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results