accessibility menu, dialog, popup

UserName

H3COBrNOO

CAS RN 18640-58-9

IUPAC Name: 1-(4-bromo-3-nitrophenyl)ethan-1-one
Synonyms: 1-(4-bromo-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 244.04
Molecular Formula: C8H6BrNO3
InChi Key: YFVOFFKNHQTQQE-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)