CAS RN 1861-32-1
CAS RN 1861-32-1
IUPAC Name:
1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate
Synonyms:
dacthal caswell no. 382 chlorthal dimethyl ester daktal dimethylester kyseliny tetrachlortereftalove chlorthal-methyl epa pesticide chemical code 078701 dac 4 tetral dacthalor
Molecular Weight (g/mol):
331.95
Molecular Formula:
C10H6Cl4O4
InChi Key:
NPOJQCVWMSKXDN-UHFFFAOYSA-N
SMILES:
COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl
1
–
1
of
1
results
1
Dimethyl Tetrachloroterephthalate 96.0+%, TCI America™
CAS: 1861-32-1 Molecular Formula: C10H6Cl4O4 Molecular Weight (g/mol): 331.95 MDL Number: MFCD00014902 InChI Key: NPOJQCVWMSKXDN-UHFFFAOYSA-N Synonym: Tetrachloroterephthalic Acid Dimethyl Ester PubChem CID: 2943 ChEBI: CHEBI:34664 IUPAC Name: 1,4-dimethyl 2,3,5,6-tetrachlorobenzene-1,4-dicarboxylate SMILES: COC(=O)C1=C(Cl)C(Cl)=C(C(=O)OC)C(Cl)=C1Cl