accessibility menu, dialog, popup

UserName

BNNNNK

CAS RN 18583-59-0

IUPAC Name: potassium bis(1H-pyrazol-1-yl)-λ¹-boranuide
Synonyms: potassium bis(pyrazol-1-yl)-λ¹-boranuide
Molecular Weight (g/mol): 184.05
Molecular Formula: C6H6BKN4
InChi Key: KIXPNQZZOUCWGN-UHFFFAOYSA-N
SMILES: [K+].[B-](N1C=CC=N1)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results