accessibility menu, dialog, popup

UserName

H3CH3COOOHOHOHO

CAS RN 18549-40-1

IUPAC Name: 1-{6-hydroxy-2,2-dimethyl-tetrahydro-2H-furo[2,3-d][1,3]dioxol-5-yl}ethane-1,2-diol
Synonyms: 1-{6-hydroxy-2,2-dimethyl-tetrahydrofuro[2,3-d][1,3]dioxol-5-yl}ethane-1,2-diol
Molecular Weight (g/mol): 220.22
Molecular Formula: C9H16O6
InChi Key: BGGCXQKYCBBHAH-UHFFFAOYNA-N
SMILES: CC1(C)OC2OC(C(O)CO)C(O)C2O1
Quantity 
  • (2)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)