accessibility menu, dialog, popup

UserName

H3CSiCH3ClCl

CAS RN 18407-07-3

IUPAC Name: dichloro(dodecyl)methylsilane
Synonyms: dichloro(dodecyl)methylsilane
Molecular Weight (g/mol): 283.35
Molecular Formula: C13H28Cl2Si
InChi Key: ZAADUWCLWTWDSZ-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC[Si](C)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results