accessibility menu, dialog, popup

UserName

H3CNHN

CAS RN 18364-47-1

IUPAC Name: N-methylpyridin-3-amine
Synonyms: N-methylpyridin-3-amine
Molecular Weight (g/mol): 108.14
Molecular Formula: C6H8N2
InChi Key: DBGFNLVRAFYZBI-UHFFFAOYSA-N
SMILES: CNC1=CC=CN=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (2)