accessibility menu, dialog, popup

UserName

H3CN

CAS RN 18300-91-9

IUPAC Name: pentadecanenitrile
Synonyms: pentadecanenitrile
Molecular Weight (g/mol): 223.4
Molecular Formula: C15H29N
InChi Key: KRKQHNVYOWTEQO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results