CAS RN 18292-38-1
IUPAC Name:
trimethyl(2-methylprop-2-en-1-yl)silane
Synonyms:
trimethyl(2-methylprop-2-en-1-yl)silane
Molecular Weight (g/mol):
128.29
Molecular Formula:
C7H16Si
InChi Key:
OWVJMAKUWHECNI-UHFFFAOYSA-N
SMILES:
CC(=C)C[Si](C)(C)C
1
–
1
of
1
results
1
Methallyltrimethylsilane 95.0+%, TCI America™
CAS: 18292-38-1 Molecular Formula: C7H16Si Molecular Weight (g/mol): 128.29 MDL Number: MFCD00054404 InChI Key: OWVJMAKUWHECNI-UHFFFAOYSA-N Synonym: Trimethyl(2-methylallyl)silane PubChem CID: 642554 IUPAC Name: trimethyl(2-methylprop-2-enyl)silane SMILES: CC(=C)C[Si](C)(C)C