accessibility menu, dialog, popup

UserName

H3CONHOH

CAS RN 18266-55-2

IUPAC Name: N-(2-hydroxyethyl)propanamide
Synonyms: N-(2-hydroxyethyl)propanamide
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: GQKLTNAIFDFUDN-UHFFFAOYSA-N
SMILES: CCC(=O)NCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results