accessibility menu, dialog, popup

UserName

CH3OSiCH3ClOH3C

CAS RN 18171-19-2

IUPAC Name: (3-chloropropyl)dimethoxymethylsilane
Synonyms: (3-chloropropyl)dimethoxymethylsilane
Molecular Weight (g/mol): 182.72
Molecular Formula: C6H15ClO2Si
InChi Key: KNTKCYKJRSMRMZ-UHFFFAOYSA-N
SMILES: CO[Si](C)(CCCCl)OC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results