accessibility menu, dialog, popup

UserName

H3COSiOCH3OH3C

CAS RN 18052-76-1

IUPAC Name: trimethoxy(naphthalen-1-yl)silane
Synonyms: trimethoxy(naphthalen-1-yl)silane
Molecular Weight (g/mol): 248.35
Molecular Formula: C13H16O3Si
InChi Key: ZSOVVFMGSCDMIF-UHFFFAOYSA-N
SMILES: CO[Si](OC)(OC)C1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results