accessibility menu, dialog, popup

UserName

H3CSiCH3H3COOSiCH3CH3H3C

CAS RN 17887-80-8

IUPAC Name: 2,2,8,8-tetramethyl-3,7-dioxa-2,8-disilanonane
Synonyms: 2,2,8,8-tetramethyl-3,7-dioxa-2,8-disilanonane
Molecular Weight (g/mol): 220.46
Molecular Formula: C9H24O2Si2
InChi Key: DWHYMKOUICVFHL-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OCCCO[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results